Learn More
TARGETMOL CHEMICALS INC CAPZIMIN 5MG
SDP
Catalog No. 502581695
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Capzimin is a selective inhibitor of proteasome isopeptidase Rpn11. purity: 98%
Catalog No. 50-258-1695 Supplier TARGETMOL CHEMICALS INC Supplier No. T106725MG
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Capzimin is a selective inhibitor of proteasome isopeptidase Rpn11. purity: 98%