Learn More
Globe Scientific™ Car Power Adapter for Portable Clinical Centrifuge
Shop All Globe Scientific ProductsDescription
- Keep your GCC-P Portable Clinical Centrifuge operating on the go with this car power adapter.
Specifications
Specifications
| For Use With (Equipment) | GCC-P Portable Clinical Centrifuge |
| Voltage | 12 VDC |
| Product Type | Power Adapter |