Learn More
Cell Signaling Technology CAR (Whitlow/218 Linker) Cell Enrichment Kit, Immunoaffinity, 1 Kit
SDP
Catalog No. 503131681
Encompass
The CAR (Whitlow/218 Linker) Cell Enrichment Kit is designed to positively select cells engineered to express a Whitlow/218 linker-containing single-chain variable fragment (scFv)-based chimeric antigen receptor (CAR) on the cell surface. This kit uses an indirect magnetic bead-based immunoaffinity enrichment protocol to yield highly pure, viable, and bead-free CAR positive cells from complex, heterogeneous cell populations.
  • Enables positive selection of cells expressing scFv-based CARs with a Whitlow/218 linker.
  • Produces highly pure, viable cells that are free of magnetic beads after processing.
  • Utilizes paramagnetic polystyrene beads (4.5 μm diameter) coupled to streptavidin via a DNase I-cleavable DNA linker.
  • Facilitates downstream analyses such as flow cytometry and single-cell omics assays.
  • Features a column-free isolation method using a magnet.
Catalog No. 50-313-1681 Supplier Cell Signaling Technology Supplier No. 65686P
The CAR (Whitlow/218 Linker) Cell Enrichment Kit is designed to positively select cells engineered to express a Whitlow/218 linker-containing single-chain variable fragment (scFv)-based chimeric antigen receptor (CAR) on the cell surface. This kit uses an indirect magnetic bead-based immunoaffinity enrichment protocol to yield highly pure, viable, and bead-free CAR positive cells from complex, heterogeneous cell populations.
  • Enables positive selection of cells expressing scFv-based CARs with a Whitlow/218 linker.
  • Produces highly pure, viable cells that are free of magnetic beads after processing.
  • Utilizes paramagnetic polystyrene beads (4.5 μm diameter) coupled to streptavidin via a DNase I-cleavable DNA linker.
  • Facilitates downstream analyses such as flow cytometry and single-cell omics assays.
  • Features a column-free isolation method using a magnet.