Learn More
Grainger Carabiner Steel 4-1/16 in L Auto-Lock
Catalog No. 502767729
Encompass_Preferred
The Condor Carabiner Double Action Twist is a high quality carabiner made from stainless steel with a maximum load limit of 3,600 lbs. It features a double action twist lock gate for added security and measures 5 inches long by 3 inches wide. This carabiner can be used for a variety of industrial, rescue, tactical, recreational and military applications including securing loads, creating anchor points, and attaching gear. It is made in the USA and comes with a limited lifetime warranty from the manufacturer.
Catalog No. 50-276-7729 Supplier Grainger Supplier No. 16V856
The Condor Carabiner Double Action Twist is a high quality carabiner made from stainless steel with a maximum load limit of 3,600 lbs. It features a double action twist lock gate for added security and measures 5 inches long by 3 inches wide. This carabiner can be used for a variety of industrial, rescue, tactical, recreational and military applications including securing loads, creating anchor points, and attaching gear. It is made in the USA and comes with a limited lifetime warranty from the manufacturer.