Learn More
Grainger Honeywell Miller Carabiner, Triple-Action Twist-Lock, Steel, 1 in. Throat Opening, 1 ea
Catalog No. 502794696
Encompass_Preferred
This steel triple-action twist-lock carabiner is designed for reliable performance in fall protection applications. It provides a secure connection with a 1 in. throat opening and robust construction for industrial use.
  • Triple-action twist-lock mechanism for enhanced security
  • Constructed from durable steel
  • 400 lbs. weight capacity for one person
  • 5,000 lbs. minimum tensile strength
  • 3,600 lbs. gate load strength
  • Certified to ANSI Z359.1, OSHA 1926.502, CSA Z259.12-01, and ANSI A10.32-2004 standards
Catalog No. 50-279-4696 Supplier Grainger Supplier No. 19Z409
This steel triple-action twist-lock carabiner is designed for reliable performance in fall protection applications. It provides a secure connection with a 1 in. throat opening and robust construction for industrial use.
  • Triple-action twist-lock mechanism for enhanced security
  • Constructed from durable steel
  • 400 lbs. weight capacity for one person
  • 5,000 lbs. minimum tensile strength
  • 3,600 lbs. gate load strength
  • Certified to ANSI Z359.1, OSHA 1926.502, CSA Z259.12-01, and ANSI A10.32-2004 standards