Learn More
Grainger Carabiner Steel 8-1/8 in L Auto-Lock
Catalog No. 502766389
Encompass_Preferred
The Condor Carabiner Double Action Twist is a high quality carabiner made from aluminum alloy with a maximum breaking strength of 3,600 lbs. It features a double action twist lock gate for added security and measures 3.15" x 2" x 0.59". This carabiner can be used for climbing, rigging, rescue, tactical, outdoor, and industrial applications. The anodized finish offers corrosion resistance. Overall, this is a strong, durable carabiner suitable for demanding situations where reliability is critical.
Catalog No. 50-276-6389 Supplier Grainger Supplier No. 16V858
The Condor Carabiner Double Action Twist is a high quality carabiner made from aluminum alloy with a maximum breaking strength of 3,600 lbs. It features a double action twist lock gate for added security and measures 3.15" x 2" x 0.59". This carabiner can be used for climbing, rigging, rescue, tactical, outdoor, and industrial applications. The anodized finish offers corrosion resistance. Overall, this is a strong, durable carabiner suitable for demanding situations where reliability is critical.