Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caraway Oil, Spectrum™ Chemical
SDP

Catalog No. 18601434 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-434 100 mL Amber Glass Bottle
18-601-435 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-601-434 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C1171100ML
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

C1171, 8000-42-8

Spectrum™ Chemical Caraway Oil , also known as meridian fennel, is a biennial plant and can be used as a spice in breads. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 8000-42-8

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 100 mL
UN Number UN1197
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.