Learn More
TARGETMOL CHEMICALS INC Carazolol 100MG
SDP
Catalog No. 502877847
Encompass_Preferred
Also available in 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Carazolol (Suacron) is a high-affinity antagonist/partial inverse agonist (also referred to as a beta blocker) of the beta-adrenergic receptor. Purity 99.08%
Catalog No. 50-287-7847 Supplier TARGETMOL CHEMICALS INC Supplier No. T3284100MG
Also available in 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Carazolol (Suacron) is a high-affinity antagonist/partial inverse agonist (also referred to as a beta blocker) of the beta-adrenergic receptor. Purity 99.08%