Learn More
Selleck Chemical LLC Carazolol
SDP
Catalog No. 5000004652
Encompass_Preferred
Carazolol is a non-specific -adrenergic receptor blocking agent and binds to receptors irreversibly
Catalog No. 50-000-04652 Supplier Selleck Chemical LLC Supplier No. S64475MG
Carazolol is a non-specific -adrenergic receptor blocking agent and binds to receptors irreversibly