Learn More
Apexbio Technology LLC CARAZOLOL-250MG
SDP
Catalog No. 5000570411
Encompass_Preferred
Carazolol (CAS No 57775-29-8) is a research compound used in biomedical sciences originating as a non-selective beta-adrenergic antagonist It primarily influences beta-adrenergic receptors by inhibiting catecholamine binding thereby affecting pathways related to cardiovascular and respiratory functions Carazolol demonstrates common in vitro activity within the nanomolar to low micromolar range reflecting its potency in various assays It is typically applied in cell models to study receptor interactions and response modulation and in animal studies for examining physiological effects on heart rate and airway dynamics Additionally it is used in drug screening to explore its potential therapeutic applications or side effects in beta-blocker development Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-70411 Supplier Apexbio Technology LLC Supplier No. C5802250
Carazolol (CAS No 57775-29-8) is a research compound used in biomedical sciences originating as a non-selective beta-adrenergic antagonist It primarily influences beta-adrenergic receptors by inhibiting catecholamine binding thereby affecting pathways related to cardiovascular and respiratory functions Carazolol demonstrates common in vitro activity within the nanomolar to low micromolar range reflecting its potency in various assays It is typically applied in cell models to study receptor interactions and response modulation and in animal studies for examining physiological effects on heart rate and airway dynamics Additionally it is used in drug screening to explore its potential therapeutic applications or side effects in beta-blocker development Experimental concentrations typically depend on the specific research design