Learn More
Medchemexpress LLC Carazolol | 57775-29-8 | 99.7% | 298.38 g/mol | C18H22N2O2 | 25 MG
SDP
Catalog No. 5000241497
Encompass_Preferred
Carazolol is a research-grade β-adrenoceptor ligand used in pharmacology and receptor-binding studies. It functions as a highly potent β1/β2 adrenoceptor antagonist and is also reported to act as a potent, selective β3-adrenoceptor agonist. The compound is supplied as a solid with reported high purity and defined molecular properties, intended for laboratory research use only.
  • Highly potent β1/β2 adrenoceptor antagonist
  • Also acts as a potent, selective β3-adrenoceptor agonist
  • Suitable for in vitro pharmacology and receptor-binding studies
  • High reported purity: 99.71%
  • Molecular formula C18H22N2O2; molecular weight 298.38 g/mol
  • Provided in small research pack sizes suitable for laboratory workflows
Catalog No. 50-002-41497 Supplier Medchemexpress LLC Supplier No. HY10732725MG
Carazolol is a research-grade β-adrenoceptor ligand used in pharmacology and receptor-binding studies. It functions as a highly potent β1/β2 adrenoceptor antagonist and is also reported to act as a potent, selective β3-adrenoceptor agonist. The compound is supplied as a solid with reported high purity and defined molecular properties, intended for laboratory research use only.
  • Highly potent β1/β2 adrenoceptor antagonist
  • Also acts as a potent, selective β3-adrenoceptor agonist
  • Suitable for in vitro pharmacology and receptor-binding studies
  • High reported purity: 99.71%
  • Molecular formula C18H22N2O2; molecular weight 298.38 g/mol
  • Provided in small research pack sizes suitable for laboratory workflows