Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carazolol | 57775-29-8 | MFCD00864588 | 99.7% | 298.38 g·mol⁻¹ | C18H22N2O2 | 1 ML
SDP

Catalog No. 5000241150
Encompass_Preferred

Carazolol is a potent β1/β2 adrenoceptor antagonist used in cardiovascular and hypertension research; it also shows activity as a selective β3 adrenoceptor agonist. It is supplied as a solid and as a ready-to-use 10 mM solution in DMSO for use in receptor pharmacology, binding assays, and cell-based studies.

  • High potency β1/β2 adrenoceptor antagonist.
  • Selective β3 adrenoceptor agonist activity.
  • Available as solid and as a 10 mM solution in DMSO.
  • High purity (99.7%) suitable for analytical and biological applications.
  • Molecular formula C18H22N2O2; molecular weight 298.38 g·mol⁻¹.

Catalog No. 50-002-41150 Supplier Medchemexpress LLC Supplier No. HY10732710MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carazolol is a potent β1/β2 adrenoceptor antagonist used in cardiovascular and hypertension research; it also shows activity as a selective β3 adrenoceptor agonist. It is supplied as a solid and as a ready-to-use 10 mM solution in DMSO for use in receptor pharmacology, binding assays, and cell-based studies.

  • High potency β1/β2 adrenoceptor antagonist.
  • Selective β3 adrenoceptor agonist activity.
  • Available as solid and as a 10 mM solution in DMSO.
  • High purity (99.7%) suitable for analytical and biological applications.
  • Molecular formula C18H22N2O2; molecular weight 298.38 g·mol⁻¹.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.