Learn More
Medchemexpress LLC Carazolol | 57775-29-8 | MFCD00864588 | 99.7% | 298.38 g/mol | C18H22N2O2 | 10 MG

Supplier: Medchemexpress LLC HY10732710MG
Carazolol is a potent, high-affinity β-adrenoceptor antagonist used as a research reagent for pharmacology and hypertension studies; it also shows selective β3-adrenoceptor agonist activity. Provided as an analytical standard, this material is intended for laboratory research use and not for human or clinical applications.
- High assay purity: 99.7%.
- Chemical formula C18H22N2O2 and molecular weight 298.38 g/mol.
- Solid, white to off-white appearance for easy handling.
- Storage: protect from light; store at 4°C; in solution store at -20°C (1 month) or -80°C (6 months).
- Suitable for receptor pharmacology, analytical reference standards, and method development.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.