Learn More
Medchemexpress LLC Carazolol | 57775-29-8 | MFCD00864588 | 99.7% | 298.38 g·mol⁻¹ | C18H22N2O2 | 50 MG
SDP
Catalog No. 5000241498
Encompass_Preferred
Carazolol is a research-grade adrenergic ligand used as a high-potency β1/β2 adrenoceptor antagonist and a potent, selective β3-adrenoceptor agonist. It is intended for use as an analytical standard and reagent in pharmacology and hypertension research. Supporting documents such as a datasheet, certificate of analysis, and safety data sheet are available.
  • High potency β1/β2 adrenoceptor antagonist for receptor studies.
  • Potent, selective β3-adrenoceptor agonist activity.
  • High purity (99.7%) suitable for analytical applications.
  • Supplied in a small research package (50 mg).
  • Supporting documents available: datasheet, COA, SDS.
Catalog No. 50-002-41498 Supplier Medchemexpress LLC Supplier No. HY10732750MG
Carazolol is a research-grade adrenergic ligand used as a high-potency β1/β2 adrenoceptor antagonist and a potent, selective β3-adrenoceptor agonist. It is intended for use as an analytical standard and reagent in pharmacology and hypertension research. Supporting documents such as a datasheet, certificate of analysis, and safety data sheet are available.
  • High potency β1/β2 adrenoceptor antagonist for receptor studies.
  • Potent, selective β3-adrenoceptor agonist activity.
  • High purity (99.7%) suitable for analytical applications.
  • Supplied in a small research package (50 mg).
  • Supporting documents available: datasheet, COA, SDS.