Learn More
Medchemexpress LLC Carazolol-d7 10mg | 1173021-02-7 | 10MG
SDP
Catalog No. 5000241131
Encompass_Preferred
Carazolol-d7 is the deuterium labeled Carazolol (HY-107327) Carazolol is a highly potent antagonist of 1/ 2 adrenoceptor Carazolol is also a potent selective 3-adrenoceptor agonist Carazolol can be used in the research of hypertension[1
Catalog No. 50-002-41131 Supplier Medchemexpress LLC Supplier No. HY107327S10MG
Carazolol-d7 is the deuterium labeled Carazolol (HY-107327) Carazolol is a highly potent antagonist of 1/ 2 adrenoceptor Carazolol is also a potent selective 3-adrenoceptor agonist Carazolol can be used in the research of hypertension[1