Learn More
Cayman Chemical Carazolol-d7 10mg
SDP
Catalog No. 502913517
Encompass_Preferred
An internal standard for the quantification of carazolol by GC- or LC-MS
Catalog No. 50-291-3517 Supplier Cayman Chemical Supplier No. 3556010 MG
An internal standard for the quantification of carazolol by GC- or LC-MS