Learn More
Medchemexpress LLC Carazolol-d7 | 1173021-02-7 | ≥98.0% | 305.4 | C18H15D7N2O2 | 25 MG
SDP
Catalog No. 5000241162
Encompass_Preferred
Carazolol-d7 is the deuterium-labeled analog of carazolol used as an internal standard and reference material for analytical and pharmacological research. It supports accurate quantification in mass spectrometry and is commonly used in metabolic and pharmacokinetic studies.
  • Deuterium-labeled internal standard for LC-MS and GC-MS.
  • High chemical purity ≥98.0%.
  • Suitable for pharmacokinetic and metabolic studies.
  • Provides reliable quantification and method validation.
  • Supplied as a stable solid reference standard.
Catalog No. 50-002-41162 Supplier Medchemexpress LLC Supplier No. HY107327S25MG
Carazolol-d7 is the deuterium-labeled analog of carazolol used as an internal standard and reference material for analytical and pharmacological research. It supports accurate quantification in mass spectrometry and is commonly used in metabolic and pharmacokinetic studies.
  • Deuterium-labeled internal standard for LC-MS and GC-MS.
  • High chemical purity ≥98.0%.
  • Suitable for pharmacokinetic and metabolic studies.
  • Provides reliable quantification and method validation.
  • Supplied as a stable solid reference standard.