Learn More
Medchemexpress LLC Carazolol hydrochloride | 51997-43-4 | MFCD03428052 | >98.0% | 334.8 g/mol | C18H23ClN2O2 | 5 MG
SDP
Catalog No. 5000284458
Encompass_Preferred
Carazolol hydrochloride is the hydrochloride salt of carazolol, a potent β1/β2-adrenoceptor antagonist used as a research reagent in cardiovascular and receptor pharmacology studies. Supplied as a solid for laboratory use.
  • Potent β1/β2-adrenoceptor antagonist for receptor pharmacology research.
  • Supplied as a hydrochloride salt for improved stability and handling.
  • High purity (>98.0%).
  • Molecular weight 334.8 g/mol; formula C18H23ClN2O2.
  • Provided in small research quantities suitable for assay development (5 mg).
Catalog No. 50-002-84458 Supplier Medchemexpress LLC Supplier No. HYW5172645MG
Carazolol hydrochloride is the hydrochloride salt of carazolol, a potent β1/β2-adrenoceptor antagonist used as a research reagent in cardiovascular and receptor pharmacology studies. Supplied as a solid for laboratory use.
  • Potent β1/β2-adrenoceptor antagonist for receptor pharmacology research.
  • Supplied as a hydrochloride salt for improved stability and handling.
  • High purity (>98.0%).
  • Molecular weight 334.8 g/mol; formula C18H23ClN2O2.
  • Provided in small research quantities suitable for assay development (5 mg).