Learn More
Medchemexpress LLC Carazolol hydrochloride | 51997-43-4 | MFCD03428052 | >98.0% | 334.84 g/mol | C18H23ClN2O2 | 10 MG
SDP
Catalog No. 5000284088
Encompass_Preferred
Carazolol hydrochloride is a research reagent used as a potent β-adrenoceptor antagonist (β1/β2) and a reported β3-adrenoceptor agonist. It is provided as a solid for use in pharmacology and receptor studies. Consult the product Certificate of Analysis for detailed purity, storage, and handling information.
  • Potent β1/β2 adrenoceptor antagonist and β3 agonist.
  • Suitable for receptor pharmacology and hypertension research.
  • Reported purity greater than 98% for typical catalog items.
  • Identified by CAS 51997-43-4 for unambiguous referencing.
  • Molecular weight 334.84 g/mol; molecular formula C18H23ClN2O2.
Catalog No. 50-002-84088 Supplier Medchemexpress LLC Supplier No. HYW51726410MG
Carazolol hydrochloride is a research reagent used as a potent β-adrenoceptor antagonist (β1/β2) and a reported β3-adrenoceptor agonist. It is provided as a solid for use in pharmacology and receptor studies. Consult the product Certificate of Analysis for detailed purity, storage, and handling information.
  • Potent β1/β2 adrenoceptor antagonist and β3 agonist.
  • Suitable for receptor pharmacology and hypertension research.
  • Reported purity greater than 98% for typical catalog items.
  • Identified by CAS 51997-43-4 for unambiguous referencing.
  • Molecular weight 334.84 g/mol; molecular formula C18H23ClN2O2.