Learn More
Medchemexpress LLC Carazolol hydrochloride | 51997-43-4 | MFCD03428052 | >98.0% | 334.84 g/mol | C18H23ClN2O2 | 25 MG
SDP
Catalog No. 5000284120
Encompass_Preferred
Carazolol hydrochloride is the hydrochloride salt of carazolol, a potent β1/β2 adrenoceptor antagonist and selective β3-adrenoceptor agonist used in cardiovascular and hypertension research. It is supplied for research and analytical applications, with supporting documentation for handling and use.
  • High affinity for β-adrenoceptors, enabling receptor binding studies.
  • Hydrochloride salt form increases solubility in aqueous solvents.
  • Available in research-scale quantities suitable for analytical assays.
  • Characterized chemical identity with CAS 51997-43-4 for unambiguous identification.
  • Datasheet and handling instructions available to support safe laboratory use.
Catalog No. 50-002-84120 Supplier Medchemexpress LLC Supplier No. HYW51726425MG
Carazolol hydrochloride is the hydrochloride salt of carazolol, a potent β1/β2 adrenoceptor antagonist and selective β3-adrenoceptor agonist used in cardiovascular and hypertension research. It is supplied for research and analytical applications, with supporting documentation for handling and use.
  • High affinity for β-adrenoceptors, enabling receptor binding studies.
  • Hydrochloride salt form increases solubility in aqueous solvents.
  • Available in research-scale quantities suitable for analytical assays.
  • Characterized chemical identity with CAS 51997-43-4 for unambiguous identification.
  • Datasheet and handling instructions available to support safe laboratory use.