Learn More
Aniara Diagnostica LLC Carbonate-Bicarbonate Buffer pH 9.6, 24 tablets (blister pack)
Catalog No. 501008801
Encompass
Medicago Carb-bicarb buffer pH 9.6 supplied as preweighed tablets yields 100 mL 0.05 M sodium carbonate-bicarbonate buffer with pH 9.6 at 25C. Preservative-free. Use as coating buffer for RIA and EIA techniques. 24-count blister pack.
Catalog No. 50-100-8801 Supplier Aniara Diagnostica LLC Supplier No. A09892224
Medicago Carb-bicarb buffer pH 9.6 supplied as preweighed tablets yields 100 mL 0.05 M sodium carbonate-bicarbonate buffer with pH 9.6 at 25C. Preservative-free. Use as coating buffer for RIA and EIA techniques. 24-count blister pack.