Learn More
MSC Mitutoyo Electronic Outside Micrometer, 0 to 1 in., Carbide Face, IP65, 1 ea
Catalog No. 5000514569
Encompass
This electronic outside micrometer features a range of 0 to 1 in. (0 to 25.40 mm) and is designed for high-precision measurement tasks. With an IP65 environmental protection rating, it is resistant to water and dust, ensuring reliable performance in various workshop environments.
  • Carbide-tipped measuring faces for enhanced wear resistance
  • Ratchet stop thimble provides tactile and audible feedback for consistent, one-handed operation
  • Includes SPC data output capability
  • Supplied with a long form calibration certificate
  • Powered by one SR44 silver oxide battery
Catalog No. 50-005-14569 Supplier MSC Supplier No. 61288320
This electronic outside micrometer features a range of 0 to 1 in. (0 to 25.40 mm) and is designed for high-precision measurement tasks. With an IP65 environmental protection rating, it is resistant to water and dust, ensuring reliable performance in various workshop environments.
  • Carbide-tipped measuring faces for enhanced wear resistance
  • Ratchet stop thimble provides tactile and audible feedback for consistent, one-handed operation
  • Includes SPC data output capability
  • Supplied with a long form calibration certificate
  • Powered by one SR44 silver oxide battery