Learn More
MSC SPI Swiss Precision Instruments Electronic Outside Micrometer, 1 in. Range, Carbide-Tipped, IP65, 1 ea.
Catalog No. 5000516630
Encompass
This deep throat electronic outside micrometer is a precision instrument designed for accurate measurements up to 1.00 mm (1 in.). It features a durable carbide-tipped measuring face and a reliable ratchet thimble for consistent pressure. Built for demanding environments, this tool maintains high accuracy and is rated IP65 against dust and liquid ingress.
  • Carbide-tipped measuring face for wear resistance
  • Deep throat design with a 12.0000 in. throat depth
  • IP65-rated protection against dust and water
  • Ratchet thimble ensures consistent measuring force
  • Rotating spindle with lever lock nut for secure measurements
  • Includes one CR2032 lithium battery
  • Calibrated to ASME 89.1.13 standards
Catalog No. 50-005-16630 Supplier MSC Supplier No. 13433685
This deep throat electronic outside micrometer is a precision instrument designed for accurate measurements up to 1.00 mm (1 in.). It features a durable carbide-tipped measuring face and a reliable ratchet thimble for consistent pressure. Built for demanding environments, this tool maintains high accuracy and is rated IP65 against dust and liquid ingress.
  • Carbide-tipped measuring face for wear resistance
  • Deep throat design with a 12.0000 in. throat depth
  • IP65-rated protection against dust and water
  • Ratchet thimble ensures consistent measuring force
  • Rotating spindle with lever lock nut for secure measurements
  • Includes one CR2032 lithium battery
  • Calibrated to ASME 89.1.13 standards