Learn More
MSC SPI Electronic Outside Micrometer, 0.00 to 1.00 mm, IP65, 1 ea
Catalog No. 5000513162
Encompass
This electronic outside micrometer is a precision instrument designed for accurate dimensional measurement. It provides reliable performance in metalworking and inspection environments.
  • Range of 0.00 to 1.00 mm
  • IP65 ingress protection rating
  • Electronic readout for precise measurements
Catalog No. 50-005-13162 Supplier MSC Supplier No. 13433305
This electronic outside micrometer is a precision instrument designed for accurate dimensional measurement. It provides reliable performance in metalworking and inspection environments.
  • Range of 0.00 to 1.00 mm
  • IP65 ingress protection rating
  • Electronic readout for precise measurements