Learn More
Carbachol, 98.3%
Description
Carbachol is a cholinergic agonist and is used for the treatment of glaucoma. It is also used as ophthalmic solution (eye drops) during ophthalmic surgery.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Carbachol is an ammonium salt and a choline ester that acts on both muscarinic and nicotinic receptors
- Carbachol activates the sympathetic nervous system; it leads to intracellular increases in Ca2+ and activates associated signaling pathways
- Carbachol is not degraded by the cholinesterases
Application
- Carbachol can inhibit apoptotic cell death, making it useful for the study of cell survival and growth in laboratory settings
- Carbachol can be used for laboratory research of any biological pathway that depends on acetylcholine, including sleep cycles and motor control
Specifications
Specifications
| Melting Point | ∽210°C (decomposition) |
| Quantity | 5 g |
| UN Number | UN2811 |
| Beilstein | 3917459 |
| Sensitivity | Hygroscopic |
| Merck Index | 14,1779 |
| Solubility Information | Soluble in water and dimethyl sulfoxide. |
| Formula Weight | 182.65 |
| Percent Purity | 98.3% |
| Chemical Name or Material | Carbachol |
Safety and Handling
| Hazard Category | H300-H500 |
| Hazard Statement | GHS H Statement H300 Fatal if swallowed. |
| Precautionary Statement | P264b-P270-P301+P310-P330-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
| EINECSNumber | 200-127-3 |
| RTECSNumber | GA0875000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.