Learn More
Abcam Carbamoylcholine chloride (Carbachol), Cholinergic agonist, 1G
Supplier:  Abcam AB1413541G
Encompass_Preferred
MW 182.65 Da. Non-selective cholinergic agonist. Cholinesterase resistant. Blocks apoptosis. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2024653