Learn More
Apexbio Technology LLC Carbadox 6804-07-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061660
Encompass_Preferred
Carbadox is an antibiotic compound employed in bacterial infection research primarily targeting pathogenic organisms associated with gastrointestinal infections in swine including swine dysentery Its antibacterial mechanism involves inhibition of bacterial DNA synthesis and disruption of nucleic acid metabolism thereby interfering with pathogen replication Carbadox is frequently utilized in microbiology studies investigating therapeutic interventions against enteric bacterial pathogens under controlled laboratory conditions In vitro studies report Carbadox demonstrating bacteriostatic activity with IC50 values typically ranging between 0 5 2 g/mL for common susceptible bacterial strains serving as a reference substance in microbial sensitivity assays
Catalog No. 50-000-61660 Supplier Apexbio Technology LLC Supplier No. A83885.1
Carbadox is an antibiotic compound employed in bacterial infection research primarily targeting pathogenic organisms associated with gastrointestinal infections in swine including swine dysentery Its antibacterial mechanism involves inhibition of bacterial DNA synthesis and disruption of nucleic acid metabolism thereby interfering with pathogen replication Carbadox is frequently utilized in microbiology studies investigating therapeutic interventions against enteric bacterial pathogens under controlled laboratory conditions In vitro studies report Carbadox demonstrating bacteriostatic activity with IC50 values typically ranging between 0 5 2 g/mL for common susceptible bacterial strains serving as a reference substance in microbial sensitivity assays