Learn More
Waters Corp Waters Carbamate Analysis Kit, EPA Method 531.2 Optimized
Catalog No. 50780056 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Carbamate Analysis Kit includes a carbamate column, HLB cartridges, vials, and reference standards. It is optimized for use with EPA Method 531.2, aiming to simplify analysis and increase confidence in results.
  • Includes carbamate column, HLB cartridges, vials, and reference standards.
  • Optimized for use with EPA Method 531.2.
  • Carbamate column designed and tested for high sensitivity and excellent resolution.
  • HLB cartridges for sample cleanup and concentration.
  • Vials are certified clean and free of background contamination.
  • Reference standards for drinking water and wastewater.
Catalog No. 50-780-056 Supplier Waters Corp Supplier No. 176001740
The Carbamate Analysis Kit includes a carbamate column, HLB cartridges, vials, and reference standards. It is optimized for use with EPA Method 531.2, aiming to simplify analysis and increase confidence in results.
  • Includes carbamate column, HLB cartridges, vials, and reference standards.
  • Optimized for use with EPA Method 531.2.
  • Carbamate column designed and tested for high sensitivity and excellent resolution.
  • HLB cartridges for sample cleanup and concentration.
  • Vials are certified clean and free of background contamination.
  • Reference standards for drinking water and wastewater.