Learn More
Pickering Laboratories Inc CARBAMATE COLUMN
SDP
Catalog No. NC1020015
Encompass_Preferred
Carbamate Column, high resolution/capacity, C8 , 4.6 x 250 mm, includes Carbamate Test Mixture 1700-0063
Catalog No. NC1020015 Supplier Pickering Laboratories Inc Supplier No. 0846250
Carbamate Column, high resolution/capacity, C8 , 4.6 x 250 mm, includes Carbamate Test Mixture 1700-0063