Learn More
Cayman Chemical Carbamazepine 10,11-epoxide, Purity: ≥98%, CAS: 36507-30-9, 5 mg
SDP
Catalog No. 502948392
Encompass_Preferred
An active metabolite of carbamazepine; has anticonvulsant activity against maximal electroshock-induced seizures in mice; has been found in wastewater effluent
Catalog No. 50-294-8392 Supplier Cayman Chemical Supplier No. 339085 MG
An active metabolite of carbamazepine; has anticonvulsant activity against maximal electroshock-induced seizures in mice; has been found in wastewater effluent