Learn More
Cayman Chemical CarbamazepIn 10 11epoxIde 10mg
SDP
Catalog No. 502949171
Encompass_Preferred
An active metabolite of carbamazepine; has anticonvulsant activity against maximal electroshock-induced seizures in mice; has been found in wastewater effluent
Catalog No. 50-294-9171 Supplier Cayman Chemical Supplier No. 3390810 MG
An active metabolite of carbamazepine; has anticonvulsant activity against maximal electroshock-induced seizures in mice; has been found in wastewater effluent