Learn More
TARGETMOL CHEMICALS INC Carbamazepine 10 11-epoxide 2
SDP
Supplier:  TARGETMOL CHEMICALS INC T3714925MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Carbamazepine 10,11-epoxide is an active metabolite of the anticonvulsant carbamazepine. It is formed by the cytochrome P450 (CYP) isoforms CYP3A4 and CYP2C8 in microsomes from HepG2 cells expressing CYP3A4 or CYP2C8, respectively. Carbamazepine 10,11-epoxide exhibits anticonvulsant activity against maximal electroshock-induced seizures in mice and has been detected in wastewater effluent [1][2][3]. Purity 98.64%
Catalog No. 50-288-0057