Learn More
Medchemexpress LLC Carbamazepine 10,11-epoxide | 36507-30-9 | 99.1% | 252.27 | 1 ML
SDP
Catalog No. 5000454281
Encompass_Preferred
Carbamazepine 10,11-epoxide is an orally active metabolite of Carbamazepine (HY-B0246). Carbamazepine has anticonvulsant effect and can be used for the research of seizures.
  • Orally active metabolite
  • Anticonvulsant effect
  • Suitable for research of seizures
Catalog No. 50-004-54281 Supplier Medchemexpress LLC Supplier No. HYW01337810MM/1ML
Carbamazepine 10,11-epoxide is an orally active metabolite of Carbamazepine (HY-B0246). Carbamazepine has anticonvulsant effect and can be used for the research of seizures.
  • Orally active metabolite
  • Anticonvulsant effect
  • Suitable for research of seizures