Learn More
Medchemexpress LLC Carbamazepine 10,11-epoxide | 36507-30-9 | 99.1% | 252.27 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW013378
Encompass_Preferred
Carbamazepine 10,11-epoxide is an orally active metabolite of Carbamazepine. This compound exhibits an anticonvulsant effect and is suitable for research related to seizures.
  • Orally active metabolite
  • Anticonvulsant effect
  • Suitable for research of seizures
Catalog No. 50-004-52150