Learn More
Medchemexpress LLC Carbamazepine 10,11-epoxide | 36507-30-9 | 99.1% | 252.27 | 5 MG
SDP
Catalog No. 5000454625
Encompass_Preferred
Carbamazepine 10,11-epoxide is an orally active metabolite of Carbamazepine. Carbamazepine has an anticonvulsant effect and can be used for the research of seizures. This product is for research use only.
  • Orally active metabolite
  • Anticonvulsant effect, relevant for seizure research
  • Appearance: Solid, off-white to light yellow color
  • Initial source: Endogenous metabolite
Catalog No. 50-004-54625 Supplier Medchemexpress LLC Supplier No. HYW0133785MG
Carbamazepine 10,11-epoxide is an orally active metabolite of Carbamazepine. Carbamazepine has an anticonvulsant effect and can be used for the research of seizures. This product is for research use only.
  • Orally active metabolite
  • Anticonvulsant effect, relevant for seizure research
  • Appearance: Solid, off-white to light yellow color
  • Initial source: Endogenous metabolite