Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbamazepine 10,11-epoxide, ≥99.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111018024 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8024 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8024 Supplier MilliporeSigma Supelco Supplier No. 9320325MG
Add to Cart
Add to Cart
Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 36507-30-9
Physical Form Neat
Linear Formula C15H12N2O2
MDL Number MFCD00467137
Synonym 1 A,10 b-Dihydro-6 H-dibenzo(b,f)oxireno[d]azepine-6-carboxamide
Type Analytical Standard
Quantity 25 mg
Formula Weight 252.27
Percent Purity ≥99.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.