Learn More
Medchemexpress LLC Carbamazepine 10,11 epoxide-d10 | 1219804-16-6 | 99.0% | 262.33 | 5 MG
SDP
Catalog No. 5000454751
Encompass_Preferred
Carbamazepine 10,11 epoxide-d10 is the deuterium labeled Carbamazepine 10,11 epoxide.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-54751 Supplier Medchemexpress LLC Supplier No. HYW013378S15MG
Carbamazepine 10,11 epoxide-d10 is the deuterium labeled Carbamazepine 10,11 epoxide.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.