Learn More
Medchemexpress LLC Carbamazepine | 298-46-4 | 99.95% | 236.27 | 100 MG
SDP
Catalog No. 5000364676
Encompass_Preferred
Carbamazepine is an orally active pressure-sensitive sodium ion channel blocker with an IC50 of 131 μM. It blocks voltage-gated Na+, Ca2+, and K+ channels and is also a HDAC inhibitor (IC50: 2 μM). Carbamazepine is an anticonvulsant and can be used for research of epilepsy and neuropathic pain.
  • Orally active pressure-sensitive sodium ion channel blocker
  • Blocks voltage-gated Na+, Ca2+, and K+ channels
  • Acts as a HDAC inhibitor
  • Anticonvulsant properties
  • Useful for research into epilepsy
  • Useful for research into neuropathic pain
Catalog No. 50-003-64676 Supplier Medchemexpress LLC Supplier No. 100MGHYB0246
Carbamazepine is an orally active pressure-sensitive sodium ion channel blocker with an IC50 of 131 μM. It blocks voltage-gated Na+, Ca2+, and K+ channels and is also a HDAC inhibitor (IC50: 2 μM). Carbamazepine is an anticonvulsant and can be used for research of epilepsy and neuropathic pain.
  • Orally active pressure-sensitive sodium ion channel blocker
  • Blocks voltage-gated Na+, Ca2+, and K+ channels
  • Acts as a HDAC inhibitor
  • Anticonvulsant properties
  • Useful for research into epilepsy
  • Useful for research into neuropathic pain