Learn More
Medchemexpress LLC Carbamazepine | 298-46-4 | MFCD00005073 | 99.95% | 1 ML
SDP
Catalog No. 5000405385
Encompass_Preferred
Carbamazepine is an orally active sodium ion channel blocker and HDAC inhibitor. It is used for research related to epilepsy and neuropathic pain, blocking voltage-gated Na+, Ca2+, and K+ channels. Its anticonvulsant properties make it suitable for various neurological studies.
  • Orally active sodium ion channel blocker
  • HDAC inhibitor with IC50 of 2 μM
  • Blocks voltage-gated Na+, Ca2+, and K+ channels
  • Used in epilepsy and neuropathic pain research
  • Anticonvulsant properties
Catalog No. 50-004-05385 Supplier Medchemexpress LLC Supplier No. HYB024610MM/1ML
Carbamazepine is an orally active sodium ion channel blocker and HDAC inhibitor. It is used for research related to epilepsy and neuropathic pain, blocking voltage-gated Na+, Ca2+, and K+ channels. Its anticonvulsant properties make it suitable for various neurological studies.
  • Orally active sodium ion channel blocker
  • HDAC inhibitor with IC50 of 2 μM
  • Blocks voltage-gated Na+, Ca2+, and K+ channels
  • Used in epilepsy and neuropathic pain research
  • Anticonvulsant properties