Learn More
Selleck Chemical LLC Carbamazepine S1693-1g
SDP
Catalog No. 503108650
Encompass_Preferred
Carbamazepine (Carbatrol NSC 169864) is a sodium channel blocker with IC50 of 131 M in rat brain synaptosomes
Catalog No. 50-310-8650 Supplier Selleck Chemical LLC Supplier No. S16931G
Carbamazepine (Carbatrol NSC 169864) is a sodium channel blocker with IC50 of 131 M in rat brain synaptosomes