Learn More
Selleck Chemical LLC Carbamazepine-50mg
SDP
Catalog No. 502389671
Encompass_Preferred
Carbamazepine (Carbatrol, NSC 169864) is a sodium channel blocker with IC50 of 131 μM in rat brain synaptosomes.
Catalog No. 50-238-9671 Supplier Selleck Chemical LLC Supplier No. S169350MG
Carbamazepine (Carbatrol, NSC 169864) is a sodium channel blocker with IC50 of 131 μM in rat brain synaptosomes.