Learn More
Apexbio Technology LLC CARBAMAZEPINE-D10-5MG
SDP
Catalog No. 5000570424
Encompass_Preferred
Carbamazepine-d10 (CAS No 132183-78-9) is a deuterated form of carbamazepine primarily used in biomedical research as a stable isotope-labeled internal standard It originates from the anticonvulsant drug carbamazepine which targets voltage-gated sodium channels inhibiting repetitive neuronal firing In vitro activities are generally observed in the nanomolar to low micromolar range making it useful for precise pharmacokinetic and bioavailability studies Typical applications include studying metabolic pathways in cell models and examining drug interactions in animal studies Additionally it assists in drug screening assays ensuring accurate quantification Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-70424 Supplier Apexbio Technology LLC Supplier No. SL10345
Carbamazepine-d10 (CAS No 132183-78-9) is a deuterated form of carbamazepine primarily used in biomedical research as a stable isotope-labeled internal standard It originates from the anticonvulsant drug carbamazepine which targets voltage-gated sodium channels inhibiting repetitive neuronal firing In vitro activities are generally observed in the nanomolar to low micromolar range making it useful for precise pharmacokinetic and bioavailability studies Typical applications include studying metabolic pathways in cell models and examining drug interactions in animal studies Additionally it assists in drug screening assays ensuring accurate quantification Experimental concentrations typically depend on the specific research design