Learn More
Medchemexpress LLC Carbamazepine-d8 | 1538624-35-9 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0246S21MG
Encompass_Preferred
Carbamazepine-d8 is the deuterium labeled Carbamazepine. It is a sodium channel blocker and an anticonvulsant agent, with an IC50 of 131 μM. This compound is for research use only and not sold to patients.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-05620