Learn More
Medchemexpress LLC Carbamazepine-d8 | 1538624-35-9 | 5 MG
SDP
Catalog No. 5000407360
Encompass_Preferred
Carbamazepine-d8 is a deuterium labeled sodium channel blocker and anticonvulsant agent. It serves as a valuable compound for quantitative analysis in various research applications.
  • Used as a tracer in research.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-07360 Supplier Medchemexpress LLC Supplier No. HYB0246S25MG
Carbamazepine-d8 is a deuterium labeled sodium channel blocker and anticonvulsant agent. It serves as a valuable compound for quantitative analysis in various research applications.
  • Used as a tracer in research.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.