Learn More
Medchemexpress LLC Carbamazepine-(Ph)d8 | 1538624-35-9 | 98.7% | 1 MG
SDP
Catalog No. 5000406159
Encompass_Preferred
Carbamazepine-(Ph)d8 is a deuterium-labeled compound intended for research use only. It can be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration, the incorporation of stable heavy isotopes into drug molecules, has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06159 Supplier Medchemexpress LLC Supplier No. HYB0246S31MG
Carbamazepine-(Ph)d8 is a deuterium-labeled compound intended for research use only. It can be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration, the incorporation of stable heavy isotopes into drug molecules, has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS