Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carbamazepine-(Ph)d8 | 298-46-4 | 98.7% | 244.32 g/mol | C15H4D8N2O | 10 MG
SDP

Catalog No. 5000297218
Encompass_Preferred

Deuterium-labeled carbamazepine (phenyl-deuterated, d8) supplied as a small-quantity analytical standard for research use only. It is intended as an internal standard for mass spectrometry, pharmacokinetic studies, and analytical method development; not for human or clinical use.

  • Suitable as an internal standard for quantitative mass spectrometry
  • High isotopic enrichment and analytical-grade purity
  • Reported purity: 98.7%
  • Molecular weight: 244.32 g/mol
  • Chemical formula: C15H4D8N2O
  • Provided as a 10 mg powder for laboratory use

Catalog No. 50-002-97218 Supplier Medchemexpress LLC Supplier No. HYB0246S310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deuterium-labeled carbamazepine (phenyl-deuterated, d8) supplied as a small-quantity analytical standard for research use only. It is intended as an internal standard for mass spectrometry, pharmacokinetic studies, and analytical method development; not for human or clinical use.

  • Suitable as an internal standard for quantitative mass spectrometry
  • High isotopic enrichment and analytical-grade purity
  • Reported purity: 98.7%
  • Molecular weight: 244.32 g/mol
  • Chemical formula: C15H4D8N2O
  • Provided as a 10 mg powder for laboratory use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.