Learn More
Medchemexpress LLC Carbamazepine (Standard) | 298-46-4 | 100.0% | 100 MG
SDP
Catalog No. 5000406917
Encompass_Preferred
Carbamazepine (Standard) is the analytical standard of Carbamazepine, intended for research and analytical applications. It is an orally active pressure-sensitive sodium ion channel blocker, blocking voltage gated Na+, Ca2+, and K+ channels, and also acts as an HDAC inhibitor. It is an anticonvulsant used for research of epilepsy and neuropathic pain, commonly utilized in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for Carbamazepine
  • Intended for research and analytical applications
  • Orally active pressure-sensitive sodium ion channel blocker
  • Blocks voltage gated Na+, Ca2+, and K+ channels
  • Acts as an HDAC inhibitor
  • Anticonvulsant for epilepsy and neuropathic pain research
  • Used in HPLC, GC, and MS experiments
Catalog No. 50-004-06917 Supplier Medchemexpress LLC Supplier No. HYB0246R100MG
Carbamazepine (Standard) is the analytical standard of Carbamazepine, intended for research and analytical applications. It is an orally active pressure-sensitive sodium ion channel blocker, blocking voltage gated Na+, Ca2+, and K+ channels, and also acts as an HDAC inhibitor. It is an anticonvulsant used for research of epilepsy and neuropathic pain, commonly utilized in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for Carbamazepine
  • Intended for research and analytical applications
  • Orally active pressure-sensitive sodium ion channel blocker
  • Blocks voltage gated Na+, Ca2+, and K+ channels
  • Acts as an HDAC inhibitor
  • Anticonvulsant for epilepsy and neuropathic pain research
  • Used in HPLC, GC, and MS experiments