Learn More
Medchemexpress LLC Carbamazepine (Standard) | 298-46-4 | 99.9% | 25 MG
SDP
Catalog No. 5000451007
Encompass_Preferred
Carbamazepine (Standard) is the analytical standard of Carbamazepine, intended for research and analytical applications. It is an orally active pressure-sensitive sodium ion channel blocker with an IC50 of 131 μM. This anticonvulsant can be used for research related to epilepsy and neuropathic pain.
  • Analytical standard of Carbamazepine
  • Orally active pressure-sensitive sodium ion channel blocker (IC50: 131 μM)
  • Blocks voltage-gated Na+, Ca2+, and K+ channels
  • Acts as an HDAC inhibitor (IC50: 2 μM)
  • Can be used for research related to epilepsy and neuropathic pain
  • Commonly used in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS)
Catalog No. 50-004-51007 Supplier Medchemexpress LLC Supplier No. 25MGHYB0246R
Carbamazepine (Standard) is the analytical standard of Carbamazepine, intended for research and analytical applications. It is an orally active pressure-sensitive sodium ion channel blocker with an IC50 of 131 μM. This anticonvulsant can be used for research related to epilepsy and neuropathic pain.
  • Analytical standard of Carbamazepine
  • Orally active pressure-sensitive sodium ion channel blocker (IC50: 131 μM)
  • Blocks voltage-gated Na+, Ca2+, and K+ channels
  • Acts as an HDAC inhibitor (IC50: 2 μM)
  • Can be used for research related to epilepsy and neuropathic pain
  • Commonly used in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS)