Learn More
Medchemexpress LLC Carbamazepine (standard) | 298-46-4 | 99.97% | 50 MG
SDP
Catalog No. 5000406158
Encompass_Preferred
Carbamazepine (Standard) is an orally active, pressure-sensitive sodium ion channel blocker, demonstrating an IC50 of 131 μM. This compound also blocks voltage-gated Na+, Ca2+, and K+ channels, and acts as a histone deacetylase (HDAC) inhibitor with an IC50 of 2 μM. It is primarily used as an anticonvulsant in research settings. This analytical standard is suitable for qualitative, quantitative, and methodological research experiments, including applications in HPLC, GC, and MS.
  • Functions as an anticonvulsant for epilepsy and neuropathic pain research.
  • Blocks voltage-gated sodium, calcium, and potassium channels.
  • Inhibits histone deacetylase (HDAC).
  • Supplied as an analytical standard for various research applications.
Catalog No. 50-004-06158 Supplier Medchemexpress LLC Supplier No. HYB0246R50MG
Carbamazepine (Standard) is an orally active, pressure-sensitive sodium ion channel blocker, demonstrating an IC50 of 131 μM. This compound also blocks voltage-gated Na+, Ca2+, and K+ channels, and acts as a histone deacetylase (HDAC) inhibitor with an IC50 of 2 μM. It is primarily used as an anticonvulsant in research settings. This analytical standard is suitable for qualitative, quantitative, and methodological research experiments, including applications in HPLC, GC, and MS.
  • Functions as an anticonvulsant for epilepsy and neuropathic pain research.
  • Blocks voltage-gated sodium, calcium, and potassium channels.
  • Inhibits histone deacetylase (HDAC).
  • Supplied as an analytical standard for various research applications.