Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbamazepine, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18601437 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-437 5 g Amber Glass Bottle
18-601-438 25 g Amber Glass Bottle
18-601-439 100 g Amber Glass Bottle
18-601-440 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-601-437 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CA2505GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

CA250, 298-46-4, C15H12N2O

Spectrum™ Chemical Carbamazepine, USP is an anti-seizure and mood-stabilizing drug. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 298-46-4
Molecular Formula C15H12N2O
Molecular Weight (g/mol) 236.27
MDL Number MFCD00005073
InChI Key FFGPTBGBLSHEPO-UHFFFAOYSA-N
IUPAC Name 2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaene-2-carboxamide
SMILES NC(=O)N1C2=CC=CC=C2C=CC2=CC=CC=C12

Specifications

CAS Max % 100%
Quantity 5 g
Percent Purity 98% to 102%
Grade USP
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.